CAS RN 2178-24-7
IUPAC Name:
ethyl 2-(2-bromophenyl)acetate
Synonyms:
ethyl 2-(2-bromophenyl)acetate
Molecular Weight (g/mol):
243.1
Molecular Formula:
C10H11BrO2
InChi Key:
MZQXAVZPEZUJIJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CC=CC=C1Br
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-bromophenylacetate, 98%
Ethyl 2-bromophenylacetate, 98%
CAS: 2178-24-7 Molecular Formula: C10H11BrO2 Molecular Weight (g/mol): 243.1 MDL Number: MFCD00051921 InChI Key: MZQXAVZPEZUJIJ-UHFFFAOYSA-N Synonym: ethyl 2-2-bromophenyl acetate,ethyl 2-bromophenylacetate,ethyl 2-bromophenyl acetate,benzeneacetic acid, 2-bromo-, ethyl ester,acmc-20al1y,2-bromo-phenyl-acetic acid ethyl ester,ethyl o-bromophenylacetate,ethyl-2-bromophenylacetate,ethyl 2-bromo-phenylacetate,ethyl 2-2-bromophenyl-acetate PubChem CID: 2780092 IUPAC Name: ethyl 2-(2-bromophenyl)acetate SMILES: CCOC(=O)CC1=CC=CC=C1Br