missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Bromoisovalerate 98.0+%, TCI America™
Supplier: TCI America E096925G
Specifications
| Ethyl 2-Bromoisovalerate | |
| Colorless | |
| C7H13BrO2 | |
| 25 g | |
| ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, 2-bromoisovaleric acid ethyl ester, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, ethyl, a-bromoisovalerate, a-bromoisovalerate | |
| CCOC(=O)C(C(C)C)Br | |
| 209.083 | |
| 209.08 | |
| Liquid |
| 609-12-1 | |
| 186°C | |
| MFCD00026855 | |
| 3265 | |
| WNFUWONOILPKNX-UHFFFAOYSA-N | |
| ethyl 2-bromo-3-methylbutanoate | |
| 79068 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 609-12-1 | |
| 209.083 | |
| WNFUWONOILPKNX-UHFFFAOYSA-N | |
| 79068 | |
| CCOC(=O)C(C(C)C)Br |
| C7H13BrO2 | |
| MFCD00026855 | |
| ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, 2-bromoisovaleric acid ethyl ester, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, ethyl, a-bromoisovalerate, a-bromoisovalerate | |
| ethyl 2-bromo-3-methylbutanoate |
Safety and Handling
EINECSNumber : (2)-3945
TSCA : Yes