missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromoisovalerate 98.0+%, TCI America™

Catalog No. p-7129253
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09695G 5 g
E096925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09695G Supplier TCI America Supplier No. E09695G

Chemical Identifiers

CAS 609-12-1
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00026855
InChI Key WNFUWONOILPKNX-UHFFFAOYSA-N
Synonym ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, ethyl 2-bromo isovalerate
PubChem CID 79068
IUPAC Name ethyl 2-bromo-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)Br

Specifications

Color Colorless
Boiling Point 186°C
Quantity 5 g
UN Number 3265
Formula Weight 209.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromoisovalerate
EINECSNumber (2)-3945
TSCA Yes