Learn More
Ethyl 2-bromoisobutyrate, 98%
CAS: 600-00-0 | C6H11BrO2 | 195.06 g/mol
$309.62 - $309.62
Chemical Identifiers
| CAS | 600-00-0 |
|---|---|
| Molecular Formula | C6H11BrO2 |
| Molecular Weight (g/mol) | 195.06 |
| MDL Number | MFCD00000123 |
| InChI Key | IOLQWGVDEFWYNP-UHFFFAOYSA-N |
| Synonym | ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester |
| PubChem CID | 11745 |
| IUPAC Name | ethyl 2-bromo-2-methylpropanoate |
| SMILES | CCOC(=O)C(C)(C)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118135000
|
Thermo Scientific Chemicals
118135000 |
500 g | Glass bottle |
Each for $309.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 600-00-0 | |
| 195.06 | |
| IOLQWGVDEFWYNP-UHFFFAOYSA-N | |
| 11745 | |
| CCOC(=O)C(C)(C)Br |
| C6H11BrO2 | |
| MFCD00000123 | |
| ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester | |
| ethyl 2-bromo-2-methylpropanoate |
Specifications
| 600-00-0 | |
| 1.3290g/mL | |
| 60°C | |
| 97.5% min. (GC) | |
| C6H11BrO2 | |
| (CH3)2C(Br)CO2C2H5 | |
| 500 g | |
| 1.329 | |
| IOLQWGVDEFWYNP-UHFFFAOYSA-N | |
| ethyl 2-bromo-2-methylpropanoate | |
| 11745 | |
| 98% | |
| Ethyl 2-bromoisobutyrate, 98% |
| Colorless to Yellow | |
| 161°C to 163°C | |
| Authentic | |
| Glass bottle | |
| 1.4430 to 1.446 | |
| MFCD00000123 | |
| 02, 296 | |
| ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester | |
| CCOC(=O)C(C)(C)Br | |
| 195.06 | |
| 195.06 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause genetic defects if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
EINECSNumber : 209-980-6
TSCA : TSCA
RUO – Research Use Only