missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Amino-4,5-dimethylthiophene-3-carboxylate 98.0+%, TCI America™
Supplier: TCI America E100525G
Specifications
| Ethyl 2-Amino-4,5-dimethylthiophene-3-carboxylate | |
| 93°C | |
| C9H13NO2S | |
| 25 g | |
| JYSDXODDWAQWJR-UHFFFAOYSA-N | |
| ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate | |
| 78536 | |
| ≥98.0% (GC,T) |
| 4815-24-1 | |
| Yellow | |
| MFCD00085051 | |
| ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate | |
| CCOC(=O)C1=C(SC(=C1C)C)N | |
| 199.268 | |
| 199.27 | |
| Crystalline Powder |
Chemical Identifiers
| 4815-24-1 | |
| 199.268 | |
| JYSDXODDWAQWJR-UHFFFAOYSA-N | |
| 78536 | |
| CCOC(=O)C1=C(SC(=C1C)C)N |
| C9H13NO2S | |
| MFCD00085051 | |
| ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate | |
| ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate |
Safety and Handling
TSCA : No