accessibility menu, dialog, popup

UserName

CAS RN 4815-24-1

IUPAC Name: ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: JYSDXODDWAQWJR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC(C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results