missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-Dihydroxy-6-methylbenzoate 98.0+%, TCI America™

Catalog No. C13165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13165G 5 g
C13161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13165G Supplier TCI America Supplier No. C13165G
Only null left

Chemical Identifiers

CAS 2524-37-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00010821
InChI Key UQSRXQMIXSZGLA-UHFFFAOYSA-N
Synonym 2-Carbethoxy-3,5-dihydroxytoluene, 4-Carbethoxy-5-methylresorcinol, 2,4-Dihydroxy-6-methylbenzoic Acid Ethyl Ester, 2-Ethoxycarbonyl-3,5-dihydroxytoluene, 4-Ethoxycarbonyl-5-methylresorcinol
PubChem CID 75653
IUPAC Name ethyl 2,4-dihydroxy-6-methylbenzoate
SMILES CCOC(=O)C1=C(C=C(C=C1C)O)O

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2,4-Dihydroxy-6-methylbenzoate