accessibility menu, dialog, popup

UserName

CAS RN 2524-37-0

H3COOCH3OHHO

CAS RN 2524-37-0

IUPAC Name: ethyl 2,4-dihydroxy-6-methylbenzoate
Synonyms: ethyl 2,4-dihydroxy-6-methylbenzoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: UQSRXQMIXSZGLA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=C(O)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results