missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-Difluorobenzoate 98.0+%, TCI America™

Catalog No. p-7129231
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09355G 5 g
E093525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09355G Supplier TCI America Supplier No. E09355G
Only null left

Chemical Identifiers

CAS 108928-00-3
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.16
MDL Number MFCD00153149
InChI Key OPQFYGPAOVCNEQ-UHFFFAOYSA-N
Synonym 2,4-difluorobenzoic acid ethyl ester, 2,4-difluoro-benzoic acid ethyl ester, pubchem3467, acmc-209w2m, ethyl-2,4-difluorobenzoate, ethyl 2,4-di-fluorobenzoate, rarechem al bi 0213, ethyl2,4-difluorobenzoate, 2,4 ethyl-difluorobenzoate, 2,4-difluorobenzoic acid, ethyl ester
PubChem CID 2737170
IUPAC Name ethyl 2,4-difluorobenzoate
SMILES CCOC(=O)C1=CC=C(F)C=C1F

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 186.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,4-Difluorobenzoate