missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,3,4,5-Tetrafluorobenzoate 98.0+%, TCI America™

Catalog No. T209425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T209425G 25 g
T2094100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T209425G Supplier TCI America Supplier No. T209425G

Chemical Identifiers

CAS 122894-73-9
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.139
MDL Number MFCD02094158
InChI Key SBHMXBYMQZRRLC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzoic acid ethyl ester, 2,3,4,5-tetrafluoro-benzoic acid ethyl ester, pubchem4072, acmc-1c5fu, ethyl2,3,4,5-tetrafluorobenzoate, 2,3,4,5-tetrafluoro-benzoicacidethylester, ethyl 2,3,4,5-tetrakis fluoranyl benzoate
PubChem CID 2774395
IUPAC Name ethyl 2,3,4,5-tetrafluorobenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1F)F)F)F

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 g
Formula Weight 222.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,3,4,5-Tetrafluorobenzoate