accessibility menu, dialog, popup

UserName

H3COOFFFF

CAS RN 122894-73-9

IUPAC Name: ethyl 2,3,4,5-tetrafluorobenzoate
Synonyms: ethyl 2,3,4,5-tetrafluorobenzoate
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: SBHMXBYMQZRRLC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results