missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 10-Bromodecanoate 96.0+%, TCI America™

Catalog No. B22655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22655G Supplier TCI America Supplier No. B22655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55099-31-5
Molecular Formula C12H23BrO2
Molecular Weight (g/mol) 279.22
MDL Number MFCD01112172
InChI Key VWHLKJQERLYMNA-UHFFFAOYSA-N
Synonym 10-Bromodecanoic Acid Ethyl Ester
PubChem CID 560452
IUPAC Name ethyl 10-bromodecanoate
SMILES CCOC(=O)CCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 279.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 10-Bromodecanoate