accessibility menu, dialog, popup

UserName

CAS RN 55099-31-5

H3COOBr

CAS RN 55099-31-5

IUPAC Name: ethyl 10-bromodecanoate
Synonyms: ethyl 10-bromodecanoate
Molecular Weight (g/mol): 279.22
Molecular Formula: C12H23BrO2
InChi Key: VWHLKJQERLYMNA-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results