missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-Methylimidazole-2-carboxylate 98.0+%, TCI America™
$48.46 - $447.38
Chemical Identifiers
| CAS | 30148-21-1 |
|---|---|
| Molecular Formula | C7H10N2O2 |
| Molecular Weight (g/mol) | 154.17 |
| MDL Number | MFCD02683326 |
| InChI Key | NOTZYDYZBOBDFE-UHFFFAOYSA-N |
| Synonym | ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester |
| PubChem CID | 4261883 |
| IUPAC Name | ethyl 1-methyl-1H-imidazole-2-carboxylate |
| SMILES | CCOC(=O)C1=NC=CN1C |
Chemical Identifiers
| 30148-21-1 | |
| 154.17 | |
| NOTZYDYZBOBDFE-UHFFFAOYSA-N | |
| 4261883 | |
| CCOC(=O)C1=NC=CN1C |
| C7H10N2O2 | |
| MFCD02683326 | |
| ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester | |
| ethyl 1-methyl-1H-imidazole-2-carboxylate |
Specifications
| 30148-21-1 | |
| White-Yellow | |
| C7H10N2O2 | |
| 1 g | |
| NOTZYDYZBOBDFE-UHFFFAOYSA-N | |
| ethyl 1-methyl-1H-imidazole-2-carboxylate | |
| 4261883 | |
| ≥98.0% (GC,T) | |
| Ethyl 1-Methylimidazole-2-carboxylate |
| 45°C | |
| 110°C | |
| MFCD02683326 | |
| ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester | |
| CCOC(=O)C1=NC=CN1C | |
| 154.17 | |
| 154.17 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No