missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Methylimidazole-2-carboxylate 98.0+%, TCI America™

Supplier:  TCI America E111425G

 View more versions of this product

Catalog No. E111425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 1-Methylimidazole-2-carboxylate
45°C
110°C
MFCD02683326
ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester
CCOC(=O)C1=NC=CN1C
154.17
154.17
Crystalline Lumps
30148-21-1
White-Yellow
C7H10N2O2
25 g
NOTZYDYZBOBDFE-UHFFFAOYSA-N
ethyl 1-methyl-1H-imidazole-2-carboxylate
4261883
≥98.0% (GC,T)

Chemical Identifiers

30148-21-1
154.17
NOTZYDYZBOBDFE-UHFFFAOYSA-N
4261883
CCOC(=O)C1=NC=CN1C
C7H10N2O2
MFCD02683326
ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester
ethyl 1-methyl-1H-imidazole-2-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions