accessibility menu, dialog, popup

UserName

CAS RN 30148-21-1

IUPAC Name: ethyl 1-methyl-1H-imidazole-2-carboxylate
Synonyms: ethyl 1-methylimidazole-2-carboxylate
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: NOTZYDYZBOBDFE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC=CN1C
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)