Learn More
Ethyl 1-bromocyclobutanecarboxylate, 96%
CAS: 35120-18-4 | C7H11BrO2 | 207.06 g/mol
$118.68 - $118.68
Chemical Identifiers
| CAS | 35120-18-4 |
|---|---|
| Molecular Formula | C7H11BrO2 |
| Molecular Weight (g/mol) | 207.06 |
| MDL Number | MFCD00001322 |
| InChI Key | UTVNSHXHFRIXMM-UHFFFAOYSA-N |
| Synonym | ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane |
| PubChem CID | 97595 |
| IUPAC Name | ethyl 1-bromocyclobutane-1-carboxylate |
| SMILES | CCOC(=O)C1(CCC1)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC188710050
|
Thermo Scientific Chemicals
188710050 |
5 g | Glass bottle |
Each for $118.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 35120-18-4 | |
| 207.06 | |
| UTVNSHXHFRIXMM-UHFFFAOYSA-N | |
| 97595 | |
| CCOC(=O)C1(CCC1)Br |
| C7H11BrO2 | |
| MFCD00001322 | |
| ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane | |
| ethyl 1-bromocyclobutane-1-carboxylate |
Specifications
| 35120-18-4 | |
| 1.2790g/mL | |
| 79°C | |
| 95% min. (GC) | |
| C7H11BrO2 | |
| MFCD00001322 | |
| 1.279 | |
| UTVNSHXHFRIXMM-UHFFFAOYSA-N | |
| ethyl 1-bromocyclobutane-1-carboxylate | |
| 97595 | |
| 96% | |
| Ethyl 1-bromocyclobutanecarboxylate |
| Colorless | |
| 85°C to 88°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| BrC4H6CO2C2H5 | |
| 5 g | |
| ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane | |
| CCOC(=O)C1(CCC1)Br | |
| 207.06 | |
| 207.06 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 252-384-6
RUO – Research Use Only