missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1,3-Dithiane-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America D16485G
Specifications
| Ethyl 1,3-Dithiane-2-carboxylate | |
| Yellow | |
| MFCD00006657 | |
| 3272 | |
| ANEDZEVDORCLPM-UHFFFAOYSA-N | |
| ethyl 1,3-dithiane-2-carboxylate | |
| 88552 | |
| ≥98.0% (GC) |
| 20462-00-4 | |
| C7H12O2S2 | |
| 5 g | |
| 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate | |
| CCOC(=O)C1SCCCS1 | |
| 192.291 | |
| 192.29 | |
| Liquid |
Chemical Identifiers
| 20462-00-4 | |
| 192.291 | |
| ANEDZEVDORCLPM-UHFFFAOYSA-N | |
| 88552 | |
| CCOC(=O)C1SCCCS1 |
| C7H12O2S2 | |
| MFCD00006657 | |
| 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate | |
| ethyl 1,3-dithiane-2-carboxylate |
Safety and Handling
TSCA : No