accessibility menu, dialog, popup

UserName

CAS RN 20462-00-4

IUPAC Name: ethyl 1,3-dithiane-2-carboxylate
Synonyms: ethyl 1,3-dithiane-2-carboxylate
Molecular Weight (g/mol): 192.29
Molecular Formula: C7H12O2S2
InChi Key: ANEDZEVDORCLPM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1SCCCS1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)