missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,3-Dithiane-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7126849
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16481G 1 g
D16485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16481G Supplier TCI America Supplier No. D16481G
Only null left

Chemical Identifiers

CAS 20462-00-4
Molecular Formula C7H12O2S2
Molecular Weight (g/mol) 192.291
MDL Number MFCD00006657
InChI Key ANEDZEVDORCLPM-UHFFFAOYSA-N
Synonym 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate
PubChem CID 88552
IUPAC Name ethyl 1,3-dithiane-2-carboxylate
SMILES CCOC(=O)C1SCCCS1

Specifications

Color Yellow
Quantity 1 g
UN Number 3272
Formula Weight 192.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,3-Dithiane-2-carboxylate