Learn More
Ethyl 1,3-dithiane-2-carboxylate, 98+%
CAS: 20462-00-4 | C7H12O2S2 | 192.291 g/mol
Supplier: Thermo Scientific Chemicals L0066314
Description
Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
| Ethyl 1,3-dithiane-2-carboxylate | |
| 19°C to 21°C | |
| 75°C to 77°C (0.2 mmHg) | |
| C7H12O2S2 | |
| MFCD00006657 | |
| UN1993 | |
| 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate | |
| ANEDZEVDORCLPM-UHFFFAOYSA-N | |
| ethyl 1,3-dithiane-2-carboxylate | |
| 88552 | |
| ≥98% |
| 20462-00-4 | |
| 1.22 | |
| 54°C (129°F) | |
| 1.54 | |
| 25 g | |
| 1424352 | |
| Slightly miscible with water. | |
| CCOC(=O)C1SCCCS1 | |
| 192.291 | |
| 192.3 |
Chemical Identifiers
| 20462-00-4 | |
| 192.291 | |
| ANEDZEVDORCLPM-UHFFFAOYSA-N | |
| 88552 | |
| CCOC(=O)C1SCCCS1 |
| C7H12O2S2 | |
| MFCD00006657 | |
| 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate | |
| ethyl 1,3-dithiane-2-carboxylate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 243-838-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only