missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1,3-dithiane-2-carboxylate, 98+%
Description
Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Melting Point | 19°C to 21°C |
| Density | 1.22 |
| Boiling Point | 75°C to 77°C (0.2 mmHg) |
| Flash Point | 54°C (129°F) |
| Refractive Index | 1.54 |
| Quantity | 5 g |
| UN Number | UN1993 |
| Beilstein | 1424352 |
| Solubility Information | Slightly miscible with water. |
| Formula Weight | 192.3 |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 243-838-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only