missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
Description
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Specifications
Specifications
| Melting Point | ∼150°C (decomposition) |
| Quantity | 25 g |
| Beilstein | 3581273 |
| Sensitivity | Hygroscopic |
| Solubility Information | It is soluble in water. |
| IUPAC Name | (2-ethoxy-2-oxoethyl)triphenylphosphanium bromide |
| Formula Weight | 429.3 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (Ethoxycarbonylmethyl)triphenylphosphonium bromide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 216-230-1 |
| RTECSNumber | TA2299500 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only