missing translation for 'onlineSavingsMsg'
Learn More

Ethionamide 98.0+%, TCI America™

Supplier:  TCI America E069525G

 View more versions of this product

Catalog No. E069525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethionamide
162°C
167°C
MFCD00057361
ethionamide, ethioniamide, ethyonomide, amidazine, etionamid, etioniamid, ethylisothiamide, ethinamide, thioamide, trecator
CCC1=NC=CC(=C1)C(=S)N
166.242
CHEBI:4885
≥98.0% (T)
536-33-4
Yellow
C8H10N2S
25 g
AEOCXXJPGCBFJA-UHFFFAOYSA-N
2-ethylpyridine-4-carbothioamide
2761171
166.24
Crystalline Powder

Chemical Identifiers

536-33-4
166.242
AEOCXXJPGCBFJA-UHFFFAOYSA-N
2761171
2-ethylpyridine-4-carbothioamide
C8H10N2S
MFCD00057361
ethionamide, ethioniamide, ethyonomide, amidazine, etionamid, etioniamid, ethylisothiamide, ethinamide, thioamide, trecator
CHEBI:4885
CCC1=NC=CC(=C1)C(=S)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0215

RTECSNumber : NS0350000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions