missing translation for 'onlineSavingsMsg'
Learn More

Ethionamide 98.0+%, TCI America™

Catalog No. p-7129107
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06955G 5 g
E069525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06955G Supplier TCI America Supplier No. E06955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536-33-4
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.242
MDL Number MFCD00057361
InChI Key AEOCXXJPGCBFJA-UHFFFAOYSA-N
Synonym ethionamide, ethioniamide, ethyonomide, amidazine, etionamid, etioniamid, ethylisothiamide, ethinamide, thioamide, trecator
PubChem CID 2761171
ChEBI CHEBI:4885
IUPAC Name 2-ethylpyridine-4-carbothioamide
SMILES CCC1=NC=CC(=C1)C(=S)N

Specifications

Melting Point 162°C
Color Yellow
Boiling Point 167°C
Quantity 5 g
Formula Weight 166.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethionamide
EINECSNumber (9)-0215
RTECSNumber NS0350000
TSCA No