missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)

CAS: 1239-45-8 | C21H20BrN3, C21H20BrN3 | 394.32 g/mol

$63.37 - $223.71

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Synonym 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6619206
View Documents Promotion Details
Thermo Scientific Chemicals
J6619206
5 g
Each for $223.71
Only null left
 
AAJ6619203
View Documents Promotion Details
Thermo Scientific Chemicals
J6619203
1 g
Each for $63.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1239-45-8
C21H20BrN3
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide

Specifications

1239-45-8
394.32
3642536
Weak
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
dry weight
98%
Red to purple
C21H20BrN3
ca. 261°
ZMMJGEGLRURXTF-UHFFFAOYSA-N
Ethidium Bromide, Electrophoresis Reagent
394.32
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-984-6

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only