missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETHE1 Monoclonal Antibody (K52048_8E6), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | ETHE1 |
|---|---|
| Clone | K52048_8E6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: RQLSQRGGSG APILLRQMFE PVSCTFTYLL GDRESREAVL IDPVLETAPR DAQLIKELGL RLLYAVNTHC HADHITGSGL LRSLLPGCQS VISRLSGAQA DLHIEDGDSI RFGRFALETR ASPGHTPGCV TFVLNDHSMA FTGDALLIRG CGRTDFQQGC AKTLYHSVHE KIFTLPGDCL IYPAHDYHGF TVSTVEEERT LNPRLTLSCE EFVKIMGNLN
ETHE1 Monoclonal antibody specifically detects ETHE1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ETHE1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 | |
| Recombinant human ETHE1 (immunogen range is 11-230). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ethe1 |
| K52048_8E6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O95571, Q9DCM0 | |
| 23474, 66071 | |
| IgG1 | |
| Protein A | |
| Antibody |