missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETHE1 Monoclonal Antibody (K52048_8E6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556052
Description
Sequence of this protein is as follows: RQLSQRGGSG APILLRQMFE PVSCTFTYLL GDRESREAVL IDPVLETAPR DAQLIKELGL RLLYAVNTHC HADHITGSGL LRSLLPGCQS VISRLSGAQA DLHIEDGDSI RFGRFALETR ASPGHTPGCV TFVLNDHSMA FTGDALLIRG CGRTDFQQGC AKTLYHSVHE KIFTLPGDCL IYPAHDYHGF TVSTVEEERT LNPRLTLSCE EFVKIMGNLN
ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.Specifications
| ETHE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O95571, Q9DCM0 | |
| Ethe1 | |
| Recombinant human ETHE1 (immunogen range is 11-230). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K52048_8E6 | |
| Unconjugated | |
| Ethe1 | |
| 0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 | |
| Mouse | |
| Protein A | |
| RUO | |
| 23474, 66071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |