missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETHE1 Monoclonal Antibody (K52048_8E6), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555672

 View more versions of this product

Catalog No. MA555672


Only null left
Explore available promotions

Description

Description

Sequence of this protein is as follows: RQLSQRGGSG APILLRQMFE PVSCTFTYLL GDRESREAVL IDPVLETAPR DAQLIKELGL RLLYAVNTHC HADHITGSGL LRSLLPGCQS VISRLSGAQA DLHIEDGDSI RFGRFALETR ASPGHTPGCV TFVLNDHSMA FTGDALLIRG CGRTDFQQGC AKTLYHSVHE KIFTLPGDCL IYPAHDYHGF TVSTVEEERT LNPRLTLSCE EFVKIMGNLN

ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.
Specifications

Specifications

ETHE1
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
O95571, Q9DCM0
Ethe1
Recombinant human ETHE1 (immunogen range is 11-230).
50 μg
Primary
Human, Mouse
Antibody
IgG1
Western Blot
K52048_8E6
Unconjugated
Ethe1
0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12
Mouse
Protein A
RUO
23474, 66071
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions