missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Estramustine phosphate sodium

Catalog No. AC466740010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466740010 1 g
AC466742500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466740010 Supplier Thermo Scientific Chemicals Supplier No. 466740010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52205-73-9
Molecular Formula C23H30Cl2NNa2O6P

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: 50 g/L
Formula Weight 564.35
InChI Key IIUMCNJTGSMNRO-VVSKJQCTSA-L
SMILES [Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O
IUPAC Name disodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate
Molecular Weight (g/mol) 564.35
Chemical Name or Material Estramustine phosphate sodium
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

May cause an allergic skin reaction.

Causes serious eye damage.

May cause respiratory irritation.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

EINECSNumber 257-735-7

RUO – Research Use Only