missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythromycin 98.0+%, TCI America™
$89.23 - $256.31
Chemical Identifiers
| CAS | 114-07-8 |
|---|---|
| Molecular Formula | C37H67NO13 |
| Molecular Weight (g/mol) | 733.94 |
| MDL Number | MFCD00084654 |
| InChI Key | ULGZDMOVFRHVEP-RWJQBGPGSA-N |
| PubChem CID | 12560 |
| ChEBI | CHEBI:42355 |
| IUPAC Name | (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione |
| SMILES | CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O |
Chemical Identifiers
| 114-07-8 | |
| 733.94 | |
| ULGZDMOVFRHVEP-RWJQBGPGSA-N | |
| CHEBI:42355 | |
| CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O |
| C37H67NO13 | |
| MFCD00084654 | |
| 12560 | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione |
Specifications
| Erythromycin | |
| 114-07-8 | |
| 5 g | |
| MFCD00084654 | |
| CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O | |
| 733.94 | |
| CHEBI:42355 | |
| ≥98.0% (T) |
| Crystalline Powder | |
| White/Yellow | |
| C37H67NO13 | |
| ULGZDMOVFRHVEP-RWJQBGPGSA-N | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione | |
| 12560 | |
| 733.94 | |
| Erythromycin |
Safety and Handling
RTECSNumber : KF4375000
TSCA : No