missing translation for 'onlineSavingsMsg'
Learn More
Learn More
erythro-5,6-Dodecanediol 98.0+%, TCI America™
$601.87 - $4,947.80
Chemical Identifiers
| CAS | 70859-32-4 |
|---|---|
| Molecular Formula | C12H26O2 |
| Molecular Weight (g/mol) | 202.338 |
| MDL Number | MFCD02093467 |
| InChI Key | COLYTQSCHUMMSR-NEPJUHHUSA-N |
| Synonym | erythro-5,6-Dihydroxydodecane |
| PubChem CID | 91659018 |
| IUPAC Name | (5R,6S)-dodecane-5,6-diol |
| SMILES | CCCCCCC(C(CCCC)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
D2719100MG
|
N/A | 100 mg |
Each for $601.87
|
|
|||||
|
D27191G
|
N/A | 1 g |
Each for $4,947.80
|
|
|||||
Chemical Identifiers
| 70859-32-4 | |
| 202.338 | |
| COLYTQSCHUMMSR-NEPJUHHUSA-N | |
| 91659018 | |
| CCCCCCC(C(CCCC)O)O |
| C12H26O2 | |
| MFCD02093467 | |
| erythro-5,6-Dihydroxydodecane | |
| (5R,6S)-dodecane-5,6-diol |
Specifications
| 70859-32-4 | |
| C12H26O2 | |
| erythro-5,6-Dihydroxydodecane | |
| CCCCCCC(C(CCCC)O)O | |
| 202.338 | |
| 202.34 | |
| Crystalline Powder |
| White | |
| MFCD02093467 | |
| COLYTQSCHUMMSR-NEPJUHHUSA-N | |
| (5R,6S)-dodecane-5,6-diol | |
| 91659018 | |
| ≥98.0% (GC) | |
| erythro-5,6-Dodecanediol |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : No