accessibility menu, dialog, popup

UserName

H3CCH3OHOH

CAS RN 70859-32-4

IUPAC Name: dodecane-5,6-diol
Synonyms: dodecane-5,6-diol
Molecular Weight (g/mol): 202.34
Molecular Formula: C12H26O2
InChi Key: COLYTQSCHUMMSR-UHFFFAOYNA-N
SMILES: CCCCCCC(O)C(O)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results