missing translation for 'onlineSavingsMsg'
Learn More

erythro-5,6-Dodecanediol 98.0+%, TCI America™

Catalog No. D27191G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27191G 1 g
D2719100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27191G Supplier TCI America Supplier No. D27191G

Chemical Identifiers

CAS 70859-32-4
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.338
MDL Number MFCD02093467
InChI Key COLYTQSCHUMMSR-NEPJUHHUSA-N
Synonym erythro-5,6-Dihydroxydodecane
PubChem CID 91659018
IUPAC Name (5R,6S)-dodecane-5,6-diol
SMILES CCCCCCC(C(CCCC)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 202.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material erythro-5,6-Dodecanediol
EINECSNumber (2)-0240
TSCA No