missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erucic Acid 85.0+%, TCI America™
Chemical Identifiers
| CAS | 112-86-7 |
|---|---|
| Molecular Formula | C22H42O2 |
| Molecular Weight (g/mol) | 338.576 |
| MDL Number | MFCD00063188 |
| InChI Key | DPUOLQHDNGRHBS-KTKRTIGZSA-N |
| Synonym | erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290 |
| PubChem CID | 5281116 |
| ChEBI | CHEBI:28792 |
| IUPAC Name | (Z)-docos-13-enoic acid |
| SMILES | CCCCCCCCC=CCCCCCCCCCCCC(=O)O |
Chemical Identifiers
| C22H42O2 | |
| MFCD00063188 | |
| erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290 | |
| CHEBI:28792 | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)O |
Specifications
| 112-86-7 | |
| C22H42O2 | |
| erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290 | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)O | |
| 338.576 | |
| CHEBI:28792 | |
| ≥85.0% (GC) | |
| Erucic Acid |
| White-Yellow | |
| MFCD00063188 | |
| DPUOLQHDNGRHBS-KTKRTIGZSA-N | |
| (Z)-docos-13-enoic acid | |
| 5281116 | |
| 338.58 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0609&(2)-0983
TSCA : Yes
Recommended Storage : Refrigerator