missing translation for 'onlineSavingsMsg'
Learn More

Erucic Acid 85.0+%, TCI America™

Catalog No. D096525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096525G 25 g
D0965100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D096525G Supplier TCI America Supplier No. D096525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 338.58
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Erucic Acid
EINECSNumber (2)-0609&(2)-0983
TSCA Yes
Recommended Storage Refrigerator