missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erucic Acid 85.0+%, TCI America™
Supplier: TCI America D0965100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| C22H42O2 | |
| MFCD00063188 | |
| erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290 | |
| CHEBI:28792 | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)O |
Specifications
| Erucic Acid | |
| White-Yellow | |
| 100 g | |
| erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290 | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)O | |
| 338.576 | |
| CHEBI:28792 | |
| ≥85.0% (GC) |
| 112-86-7 | |
| C22H42O2 | |
| MFCD00063188 | |
| DPUOLQHDNGRHBS-KTKRTIGZSA-N | |
| (Z)-docos-13-enoic acid | |
| 5281116 | |
| 338.58 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0609&(2)-0983
TSCA : Yes
Recommended Storage : Refrigerator