missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERM (LRRKtide) Substrate

Catalog No. PV5093
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5093 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5093 Supplier Invitrogen™ Supplier No. PV5093

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ERM (LRRKtide) Substrate

A peptide substrate based upon ezrin/radixin/moesin (ERM) that is phosphorylated by LRRK2 and LRRK2 mutants. For use with the Adapta™ Universal Kinase Assay (PV5099).

Specifications

Amino Acid Sequence RLGRDKYKTLRQIRQ
Assay Entry Biochemical ADP accumulation
Content And Storage 2 mM ERM (LRRKtide) peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA. Store at -20°C.
Detection Method Fluorescence
Quantity 1 mL
Substrate peptide based upon ezrin/radixin/moesin (ERM)
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 120892
Molecular Weight (g/mol) 1931.2 Da
Shipping Condition Dry Ice
Target Entry LRRK2 G2019S, LRRK2
Validated Kinases LRRK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.