missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Blue Black R Indicator, 0.2% (w/w) in Sodium Chloride, Ricca Chemical
$149.18 - $595.54
Chemical Identifiers
| CAS | 2538-85-4 |
|---|---|
| Molecular Formula | C20H13N2NaO5S |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | YJUGDMOGPJJWDG-UHFFFAOYSA-M |
| PubChem CID | 23702480 |
| IUPAC Name | sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate |
| SMILES | C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2902-100
|
Ricca Chemical Company
2902100 |
100 g | White Poly Wide Mouth Bottle |
Each for $149.18
|
|
||||
|
R2902000500
|
Ricca Chemical Company
R2902000500A6 |
500 g | Natural Poly Bottle |
Each for $595.54
|
|
||||
Chemical Identifiers
| 2538-85-4 | |
| Mixture | |
| 23702480 | |
| C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] |
| C20H13N2NaO5S | |
| YJUGDMOGPJJWDG-UHFFFAOYSA-M | |
| sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate |
Specifications
| 2538-85-4 , 7647-14-5 | |
| C20H13N2NaO5S | |
| C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] | |
| Mixture | |
| Indicator | |
| Sodium Chloride | |
| 100 g | |
| Eriochrome Blue Black R Indicator |
| 0.2% (w/w) | |
| YJUGDMOGPJJWDG-UHFFFAOYSA-M | |
| sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate | |
| 23702480 | |
| White Poly Wide Mouth Bottle | |
| Purple/Violet | |
| Powder |