missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Blue Black R Indicator, 0.2% (w/w) in Sodium Chloride, Ricca Chemical
Specifications
Specifications
| Concentration | 0.2% (w/w) |
| Grade | Indicator |
| Packaging | White Poly Wide Mouth Bottle |
| Solvent or Matrix | Sodium Chloride |
| Color | Purple/Violet |
| Quantity | 100 g |
| Physical Form | Powder |
| Chemical Name or Material | Eriochrome Blue Black R Indicator |