missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagents

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CE2550001C

 View more versions of this product

Catalog No. 01335323


Only null left

Description

Description

Specifications

Chemical Identifiers

123°C
Water hardness indicator
Reagent
0.97
Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol
109-86-4 , 1787-61-7
0.5%
Glass Bottle
Liquid

Specifications

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol
109-86-4 , 1787-61-7
0.5%
Glass Bottle
1 L
123°C
Water hardness indicator
Reagent
0.97
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions