missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T, ACS Reagent (EBT; Mordant Black 11), Reagents
Eriochrome Black T, ACS Reagent (EBT; Mordant Black 11), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$74.50 - $463.39
Chemical Identifiers
| CAS | 1787-61-7 |
|---|---|
| Molecular Formula | C20H12N3NaO7S |
| Molecular Weight (g/mol) | 461.38 |
| InChI Key | JHUJLRKQZAPSDP-GXTSIBQPSA-M |
| Synonym | Water hardness indicator |
| IUPAC Name | sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
| SMILES | [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
Chemical Identifiers
| 1787-61-7 | |
| 461.38 | |
| Water hardness indicator | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
| C20H12N3NaO7S | |
| JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
| sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
Specifications
| 1787-61-7 | |
| C20H12N3NaO7S | |
| JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
| sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate | |
| ACS | |
| 1.109 | |
| Solid |
| 1413°C | |
| Water hardness indicator | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O | |
| 461.38 | |
| Glass Bottle | |
| Black to Brown | |
| Eriochrome Black T, ACS Reagent (EBT, Mordant Black 11) |