missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T, ACS Reagent (EBT; Mordant Black 11), Reagents
Specifications
Specifications
| Boiling Point | 1413°C |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 1.109 |
| Color | Black to Brown |
| Quantity | 100 g |
| Physical Form | Solid |
| Chemical Name or Material | Eriochrome Black T, ACS Reagent (EBT, Mordant Black 11) |