missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T 0.5% in Triethanolamine, Reagents
Eriochrome Black T 0.5% in Triethanolamine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$188.71 - $473.29
Chemical Identifiers
| CAS | 102-71-6 , 1787-61-7 |
|---|---|
| Molecular Formula | C6H15NO3 |
| Molecular Weight (g/mol) | 149.19 |
| InChI Key | GSEJCLTVZPLZKY-UHFFFAOYSA-N |
| Synonym | Water hardness indicator |
| IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethan-1-ol |
| SMILES | OCCN(CCO)CCO |
Chemical Identifiers
| C6H15NO3 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol |
Specifications
| 335°C | |
| C6H15NO3 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol | |
| 0.5% | |
| Poly Bottle | |
| Purple | |
| Liquid |