missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T 0.5% in Triethanolamine, Reagents
Eriochrome Black T 0.5% in Triethanolamine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CE255400500A
| Quantity | 500 mL |
|---|
Chemical Identifiers
| C6H15NO3 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol |
Specifications
| Eriochrome Black T 0.5% in Triethanolamine | |
| 102-71-6 , 1787-61-7 | |
| Water hardness indicator | |
| OCCN(CCO)CCO | |
| 149.19 | |
| Reagent | |
| 1.1242 | |
| 10.5 | |
| Liquid |
| 335°C | |
| C6H15NO3 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol | |
| 0.5% | |
| Poly Bottle | |
| Purple | |
| 500 mL |