missing translation for 'onlineSavingsMsg'
Learn More

Eprosartan Mesylate 98.0+%, TCI America™

Supplier:  TCI America E11611G

 View more versions of this product

Catalog No. E11611G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Eprosartan Mesylate
248°C
C24H28N2O7S2
1 g
DJSLTDBPKHORNY-XMMWENQYSA-N
4-({2-butyl-5-[(1E)-2-carboxy-2-[(thiophen-2-yl)methyl]eth-1-en-1-yl]-1H-imidazol-1-yl}methyl)benzoic acid; methanesulfonic acid
5282474
520.62
Crystalline Powder
144143-96-4
White
MFCD08141807
eprosartan mesylate, teveten, eprosartan methanesulfonate, eprosartan mesilate, unii-8n2l1nx8s3, eprosartan mesylate usan:ban, e-2-butyl-1-p-carboxybenzyl-alpha-2-thenylimidazole-5-acrylic acid, monomethanesulfonate, teveten tn, dsstox_cid_24217
CS(O)(=O)=O.CCCCC1=NC=C(\C=C(/CC2=CC=CS2)C(O)=O)N1CC1=CC=C(C=C1)C(O)=O
520.62
CHEBI:48409
≥98.0% (HPLC,T)

Chemical Identifiers

144143-96-4
520.62
DJSLTDBPKHORNY-XMMWENQYSA-N
5282474
4-({2-butyl-5-[(1E)-2-carboxy-2-[(thiophen-2-yl)methyl]eth-1-en-1-yl]-1H-imidazol-1-yl}methyl)benzoic acid; methanesulfonic acid
C24H28N2O7S2
MFCD08141807
eprosartan mesylate, teveten, eprosartan methanesulfonate, eprosartan mesilate, unii-8n2l1nx8s3, eprosartan mesylate usan:ban, e-2-butyl-1-p-carboxybenzyl-alpha-2-thenylimidazole-5-acrylic acid, monomethanesulfonate, teveten tn, dsstox_cid_24217
CHEBI:48409
CS(O)(=O)=O.CCCCC1=NC=C(\C=C(/CC2=CC=CS2)C(O)=O)N1CC1=CC=C(C=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XN0282400

TSCA : No

SDS
Documents

Documents

Promotions

Promotions