accessibility menu, dialog, popup

UserName

CAS RN 144143-96-4

IUPAC Name: 4-({2-butyl-5-[(1E)-2-carboxy-2-[(thiophen-2-yl)methyl]eth-1-en-1-yl]-1H-imidazol-1-yl}methyl)benzoic acid; methanesulfonic acid
Synonyms: teveten eprosartan mesylate eprosartan methanesulfonate
Molecular Weight (g/mol): 520.62
Molecular Formula: C24H28N2O7S2
InChi Key: DJSLTDBPKHORNY-XMMWENQYSA-N
SMILES: CS(O)(=O)=O.CCCCC1=NC=C(\C=C(/CC2=CC=CS2)C(O)=O)N1CC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results