missing translation for 'onlineSavingsMsg'
Learn More

Eprosartan Mesylate 98.0+%, TCI America™

Catalog No. p-7129371
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1161200MG 200 mg
E11611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1161200MG Supplier TCI America Supplier No. E1161200MG
Only null left

Chemical Identifiers

CAS 144143-96-4
Molecular Formula C24H28N2O7S2
Molecular Weight (g/mol) 520.62
MDL Number MFCD08141807
InChI Key DJSLTDBPKHORNY-XMMWENQYSA-N
Synonym eprosartan mesylate, teveten, eprosartan methanesulfonate, eprosartan mesilate, unii-8n2l1nx8s3, eprosartan mesylate usan:ban, e-2-butyl-1-p-carboxybenzyl-alpha-2-thenylimidazole-5-acrylic acid, monomethanesulfonate, teveten tn, dsstox_cid_24217
PubChem CID 5282474
ChEBI CHEBI:48409
IUPAC Name 4-({2-butyl-5-[(1E)-2-carboxy-2-[(thiophen-2-yl)methyl]eth-1-en-1-yl]-1H-imidazol-1-yl}methyl)benzoic acid; methanesulfonic acid
SMILES CS(O)(=O)=O.CCCCC1=NC=C(\C=C(/CC2=CC=CS2)C(O)=O)N1CC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 248°C
Color White
Quantity 200 mg
Formula Weight 520.62
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Eprosartan Mesylate
RTECSNumber XN0282400
TSCA No