missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 99%, for biochemistry

Catalog No. AC172605000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172605000 500 g Glass bottle
AC172601000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172605000 Supplier Thermo Scientific Chemicals Supplier No. 172605000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Absorbance 0.1 max. at 260nm
Absorbance Conditions (0.1M aq. soln.) (1 cm cell)
Color White
pH 5 to 6 (10% w/v soln. at 20°C)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 252.33
Percent Purity ≥98.5%
Grade Biochemical
Chemical Name or Material EPPS
Show More Show Less

RUO – Research Use Only