missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.0

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

$181.82 - $291.27

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61296AE
View Documents
Thermo Scientific Chemicals
J61296AE
100 mL
Each for $181.82
Only null left
 
AAJ61296AK
View Documents
Thermo Scientific Chemicals
J61296AK
250 mL
Each for $291.27
Only null left
 
Description

Description

EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
HEPPS
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

0.2 M
Liquid
C9H20N2O4S
HEPPS
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
EPPS buffer soln.
8.0
100 mL
MFCD00006160
It is soluble in water.
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only